4-Ethylbenzoic acid 98%

Supplier: Ambeed

A193287-100G A193287-10G A193287-25G A193287-1000G
77029-886EA 32 USD
77029-886 77029-888 77085-032 77871-965
4-Ethylbenzoic acid 98%
4-Ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD00002570
CAS Number: 619-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR