(S)-Ethyl-2-amino-2-phenylacetate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Phg-OEt.HCl, H-Phg-OEt·HCl

A193240-100G A193240-1G A193240-5G
76986-182EA 99.73 USD
76986-182 76986-184 76986-186
(S)-Ethyl-2-amino-2-phenylacetate hydrochloride 98%
(S)-Ethyl-2-amino-2-phenylacetate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MW: 215.677 g/mol
MDL Number: MFCD02259533
CAS Number: 59410-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR