(2-Amino-3-methoxyphenyl)methanol 95%

Supplier: Ambeed
Warning

A193145-1G A193145-250MG
76986-178EA 119 USD
76986-178 76986-180
(2-Amino-3-methoxyphenyl)methanol 95%
(2-Amino-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.17 g/mol
MDL Number: MFCD09029933
CAS Number: 205877-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR