(2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-mercaptotetrahydro-2H-pyran-3,4,5-triyl triacetate 95%

Supplier: Ambeed
Warning

A193111-100MG A193111-250MG
76986-174EA 29 USD
76986-174 76986-176
(2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-mercaptotetrahydro-2H-pyran-3,4,5-triyl triacetate 95%
(2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-mercaptotetrahydro-2H-pyran-3,4,5-triyl triacetate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₉S
MDL Number: MFCD01076185
CAS Number: 50615-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR