6-(Trifluoromethyl)imidazo[2,1-b]thiazole 97%

Supplier: Ambeed
Warning

A193105-100MG A193105-1G A193105-250MG A193105-5G
76986-166EA 16.98 USD
76986-166 76986-168 76986-170 76986-172
6-(Trifluoromethyl)imidazo[2,1-b]thiazole 97%
6-(Trifluoromethyl)imidazo[2,1-b]thiazole

Precaución: for Research Use Only

Formula: C₆H₃F₃N₂S
MW: 192.16 g/mol
MDL Number: MFCD12828656
CAS Number: 109113-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR