(3-Amino-4-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Aminopyridine-4-methanol, (3-Amino-pyridin-4-yl)-methanol, 3-Amino-4-hydroxymethylpyridine

A193046-1G A193046-250MG
77012-420EA 197.33 USD
77012-420 77012-422
(3-Amino-4-pyridinyl)methanol 97%
(3-Amino-4-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD06658330
CAS Number: 152398-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR