5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
7-Bromo-6-chloro-7-deazapurine
A193005-100G
A193005-1G
A193005-25G
A193005-5G
A193005-10G
76999-036EA
157.02
USD
76999-036
76999-038
76999-040
76999-042
77600-564
5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine 97%
5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₃BrClN₃ MW: 232.47 g/mol |
MDL Number:
MFCD09702029 CAS Number: 22276-95-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...