Octahydro-2H-benzimidazole-2-thione 97%

Supplier: Ambeed
Warning

A192791-250MG A192791-5G
76999-032EA 8 USD
76999-032 77881-429
Octahydro-2H-benzimidazole-2-thione 97%
Octahydro-2H-benzimidazole-2-thione

Precaución: for Research Use Only

Formula: C₇H₁₂N₂S
MDL Number: MFCD00123649
CAS Number: 30086-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR