Bis(triphenylphosphine)nickel(II) bromide 98%
Supplier: Ambeed
|
Warning
|
A192787-10G
A192787-1G
A192787-25G
A192787-5G
A192787-100G
A192787-500G
76999-000EA
11
USD
76999-000
76999-026
76999-028
76999-030
77600-558
77881-428
Bis(triphenylphosphine)nickel(II) bromide 98%
Bis(triphenylphosphine)nickel(II) bromide
Precaución: for Research Use Only
Formula:
C₃₆H₃₀Br₂NiP₂ |
MDL Number:
MFCD00015865 CAS Number: 14126-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...