Methyl 5-(3-nitrophenyl)isoxazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A192726-100MG A192726-1G
77012-412EA 47.75 USD
77012-412 77012-414
Methyl 5-(3-nitrophenyl)isoxazole-3-carboxylate 97%
Methyl 5-(3-nitrophenyl)isoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₅
MW: 248.19 g/mol
MDL Number: MFCD03778971
CAS Number: 517870-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR