Methyl-2-amino-2-methylpropanoate hydrochloride 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Methyl alpha-aminoisobutyrate hydrochloride, H-Aib-OMe.HCl, Methyl 2-aminoisobutyrate hydrochloride, H-Aib-OMe·HCl

A192709-10G A192709-1G A192709-5G A192709-100G A192709-1000G A192709-25G A192709-500G
76986-150EA 10.61 USD
76986-150 76986-152 76986-154 77550-242 77611-844 77611-846 77683-514
Methyl-2-amino-2-methylpropanoate hydrochloride 97%, Ambeed.Inc
Methyl-2-amino-2-methylpropanoate hydrochloride

Precaución: For research use only.

Formula: C₅H₁₁NO₂·HCl
MW: 153.6 g/mol
MDL Number: MFCD00214247
CAS Number: 15028-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR