2-Ethoxy-6-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A192619-25G A192619-5G
77029-862EA 80 USD
77029-862 77029-864
2-Ethoxy-6-fluorophenylboronic acid 98%
2-Ethoxy-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MDL Number: MFCD05664322
CAS Number: 957062-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR