3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid

A192568-250MG A192568-5G
76998-996EA 14.86 USD
76998-996 76998-998
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 97%
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
MDL Number: MFCD02090708
CAS Number: 284493-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR