1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-((2,4,6-Triisopropylphenyl)sulphonyl)-1H-imidazole

A192284-1G A192284-5G
77029-858EA 25 USD
77029-858 77029-860
1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole 97%
1-(2,4,6-Triisopropylbenzenesulfonyl)imidazole

Precaución: for Research Use Only

Formula: C₁₈H₂₆N₂O₂S
MDL Number: MFCD00005283
CAS Number: 50257-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR