8-Aminoimidazo[1,2-a]pyridine 96%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Imidazo[1,2-a]pyridin-8-ylamine

A192196-10G A192196-250MG A192196-5G A192196-1G A192196-25G
77012-388EA 343.74 USD
77012-388 77012-390 77012-392 77593-228 77683-508
8-Aminoimidazo[1,2-a]pyridine 96%, Ambeed.Inc
8-Aminoimidazo[1,2-a]pyridine

Precaución: For research use only.

Formula: C₇H₇N₃
MW: 133.1500 g/mol
MDL Number: MFCD06739233
CAS Number: 73221-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR