2,6-Diamino-9,10-anthraquinone 98%

Supplier: Ambeed
Warning

A192195-1G A192195-5G
76998-986EA 6 USD
76998-986 76998-988
2,6-Diamino-9,10-anthraquinone 98%
2,6-Diamino-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O₂
MDL Number: MFCD00001234
CAS Number: 131-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR