tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-3-carboxylate 98%

Supplier: Ambeed
Warning

A192185-100MG A192185-1G A192185-250MG A192185-5G A192185-25G
76986-134EA 9 USD
76986-134 76986-136 76986-138 77550-240 77890-179
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-3-carboxylate 98%
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MDL Number: MFCD17016735
CAS Number: 1251017-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR