Fmoc-Val-Cit-PAB-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Val-Cit-PAB-OH, (9H-Fluoren-9-yl)methyl [(S)-1-[[(S)-1-[[4-(Hydroxymethyl)phenyl]amino]-1-oxo-5-ureidopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]carbamate

A192152-10MG A192152-1MG A192152-25MG A192152-5MG A192152-5G A192152-1G A192152-25G A192152-250MG A192152-100G A192152-100MG
76986-126EA 8.49 USD
76986-126 76986-128 76986-130 76986-132 77550-238 77567-790 77611-842 77611-840 77683-506 77683-507
Fmoc-Val-Cit-PAB-OH 98%
Fmoc-Val-Cit-PAB-OH

Precaución: For research use only.

Formula: C₃₃H₃₉N₅O₆
MW: 601.70 g/mol
MDL Number: MFCD22417106
CAS Number: 159858-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR