N-(2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-2-methylpropan-2-amine 95%

Supplier: Ambeed
Warning

A192135-1G A192135-250MG A192135-5G
77012-382EA 153.84 USD
77012-382 77012-384 77012-386
N-(2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-2-methylpropan-2-amine 95%
N-(2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-2-methylpropan-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₂₇BFNO₂
MW: 307.21 g/mol
MDL Number: MFCD18087733
CAS Number: 1256360-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR