3-Bromo-5-chloro-2-methoxypyridine 95%
Supplier: Ambeed
|
Warning
|
A191983-100G
A191983-10G
A191983-1G
A191983-25G
A191983-5G
77012-372EA
185.66
USD
77012-372
77012-374
77012-376
77012-378
77012-380
3-Bromo-5-chloro-2-methoxypyridine 95%
3-Bromo-5-chloro-2-methoxypyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrClNO |
MDL Number:
MFCD08059322 CAS Number: 102830-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...