Ethyl 2,5-dioxopyrrole-1-carboxylate 97%

Supplier: Ambeed
Warning

A191928-10G A191928-1G A191928-250MG
77012-366EA 103 USD
77012-366 77012-368 77012-370
Ethyl 2,5-dioxopyrrole-1-carboxylate 97%
Ethyl 2,5-dioxopyrrole-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇NO₄
MDL Number: MFCD16619570
CAS Number: 55750-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR