n-Octyl-β-D-Glucopyranoside tetraacetate 98+%

Supplier: Ambeed
Warning

Synonyms: β-​D-​Glucopyranoside, octyl, 2,​3,​4,​6-​tetraacetate, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate

A191794-1G A191794-250MG A191794-5G
76986-116EA 66.84 USD
76986-116 76986-118 76986-120
n-Octyl-β-D-Glucopyranoside tetraacetate 98+%
n-Octyl-β-D-Glucopyranoside tetraacetate

Precaución: for Research Use Only

Formula: C₂₂H₃₆O₁₀
MW: 460.515 g/mol
MDL Number: MFCD00192368
CAS Number: 38954-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR