6,7-Dimethoxy-2-methylquinazolin-4-ol 95%
Supplier: Ambeed
|
Warning
|
A191768-100MG
A191768-1G
A191768-250MG
A191768-5G
77029-832EA
8
USD
77029-832
77029-834
77029-836
77029-838
6,7-Dimethoxy-2-methylquinazolin-4-ol 95%
6,7-Dimethoxy-2-methylquinazolin-4-ol
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂O₃ |
MDL Number:
MFCD11110218 CAS Number: 35241-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...