4-Fluoroisophthalonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluorobenzene-1,3-dicarbonitrile

A191756-250MG A191756-5G
76998-976EA 30.77 USD
76998-976 76998-978
4-Fluoroisophthalonitrile 98%
4-Fluoroisophthalonitrile

Precaución: for Research Use Only

Formula: C₈H₃FN₂
MW: 146.121 g/mol
MDL Number: MFCD01657003
CAS Number: 13519-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR