4-Nitro-1H-pyrazole 98%

Supplier: Ambeed
Warning

A191546-1000G A191546-100G A191546-10G A191546-500G A191546-5G A191546-25G
77012-354EA 147 USD
77012-354 77012-356 77012-358 77012-360 77012-362 77084-824
4-Nitro-1H-pyrazole 98%
4-Nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₃N₃O₂
MDL Number: MFCD00159626
CAS Number: 2075-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR