4,5-Dimethyl-1,3-dioxol-2-one 98%
Supplier: Ambeed
|
Warning
|
A191528-100G
A191528-10G
A191528-500G
A191528-5G
A191528-25G
77012-340EA
20.16
USD
77012-340
77012-342
77012-344
77012-346
77084-820
4,5-Dimethyl-1,3-dioxol-2-one 98%
4,5-Dimethyl-1,3-dioxol-2-one
Precaución: for Research Use Only
Formula:
C₅H₆O₃ MW: 114.0993 g/mol |
MDL Number:
MFCD00135139 CAS Number: 37830-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...