Fmoc-D-CYS(tBu)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Cys(tBu)-OH

A191469-1G A191469-5G
76998-970EA 31.83 USD
76998-970 76998-972
Fmoc-D-CYS(tBu)-OH 97%
Fmoc-D-CYS(tBu)-OH

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₄S
MW: 399.505 g/mol
MDL Number: MFCD00077052
CAS Number: 131766-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR