6-Bromo-2,3-dihydrobenzofuran-3-amine hydrochloride 97%

Supplier: Ambeed
Warning

A191435-100MG A191435-250MG
77029-826EA 40 USD
77029-826 77029-828
6-Bromo-2,3-dihydrobenzofuran-3-amine hydrochloride 97%
6-Bromo-2,3-dihydrobenzofuran-3-amine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉BrClNO
MDL Number: MFCD09878723
CAS Number: 1215339-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR