4-Benzyloxy-2,6-difluorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-(Benzyloxy)-2,6-difluorophenyl)boronic acid

A191434-10G A191434-1G
77029-822EA 201 USD
77029-822 77029-824
4-Benzyloxy-2,6-difluorophenylboronic acid 95%
4-Benzyloxy-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
MDL Number: MFCD12026734
CAS Number: 156635-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR