2-(4-(Aminomethyl)piperidin-1-yl)ethanol 97%

Supplier: Ambeed
Warning

A191390-1G A191390-250MG A191390-5G
77029-816EA 101 USD
77029-816 77029-818 77029-820
2-(4-(Aminomethyl)piperidin-1-yl)ethanol 97%
2-(4-(Aminomethyl)piperidin-1-yl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O
MW: 158.25 g/mol
MDL Number: MFCD06637707
CAS Number: 21168-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR