4-(tert-Butyldimethylsilyloxy)cyclohexanone 98%

Supplier: Ambeed
Warning

Synonyms: 4-((tert-Butyldimethylsilyl)oxy)cyclohexanone

A191351-1G A191351-250MG A191351-25G A191351-5G
77029-808EA 33 USD
77029-808 77029-810 77029-812 77029-814
4-(tert-Butyldimethylsilyloxy)cyclohexanone 98%
4-(tert-Butyldimethylsilyloxy)cyclohexanone

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₂Si
MW: 228.407 g/mol
MDL Number: MFCD06411307
CAS Number: 55145-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR