4-Bromo-N,N-dimethyl-3-nitroaniline 97%

Supplier: Ambeed
Warning

A190935-100MG A190935-250MG
76986-104EA 35.01 USD
76986-104 76986-106
4-Bromo-N,N-dimethyl-3-nitroaniline 97%
4-Bromo-N,N-dimethyl-3-nitroaniline

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O₂
MW: 245.07 g/mol
MDL Number: MFCD02062996
CAS Number: 70076-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR