Ethyl 5-bromo-3-formyl-1H-indole-2-carboxylate 98%

Supplier: Ambeed
Warning

A190769-1G A190769-250MG A190769-5G
77012-326EA 154.9 USD
77012-326 77012-328 77012-330
Ethyl 5-bromo-3-formyl-1H-indole-2-carboxylate 98%
Ethyl 5-bromo-3-formyl-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₃
MW: 296.11 g/mol
MDL Number: MFCD02257704
CAS Number: 100123-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR