Pyridin-2-ylmethyl 2-(4-isobutylphenyl)propanoate 95%

Supplier: Ambeed
Warning

Synonyms: Ibuprofen piconol

A190685-10G A190685-1G A190685-5G
77012-314EA 617.45 USD
77012-314 77012-316 77012-318
Pyridin-2-ylmethyl 2-(4-isobutylphenyl)propanoate 95%
Pyridin-2-ylmethyl 2-(4-isobutylphenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₉H₂₃NO₂
MW: 297.39 g/mol
MDL Number: MFCD00866018
CAS Number: 64622-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR