(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(4-(5-Bromo-2-chlorobenzyl)phenoxy)tetrahydrofuran

A190671-100G A190671-10G A190671-1G A190671-25G A190671-5G
77012-304EA 314 USD
77012-304 77012-306 77012-308 77012-310 77012-312
(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran 97%
(S)-3-[4-(5-Bromo-2-chlorobenzyl)phenoxy]tetrahydrofuran

Precaución: for Research Use Only

Formula: C₁₇H₁₆BrClO₂
MW: 367.665 g/mol
MDL Number: MFCD27920793
CAS Number: 915095-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR