3-Ethyl-1-methyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A190473-100MG A190473-250MG
76998-958EA 39.26 USD
76998-958 76998-960
3-Ethyl-1-methyl-1H-pyrazole-5-carboxylic acid 97%
3-Ethyl-1-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD03086079
CAS Number: 26308-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR