1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,5-Tri-O-benzoyl-α-D-ribofuranose, 1,3,5-Tri-O-benzoyl-a-D-ribofuranose

A190455-100G A190455-10G A190455-1G A190455-250MG A190455-25G A190455-5G A190455-500G
76998-946EA 175 USD
76998-946 76998-948 76998-950 76998-952 76998-954 76998-956 77671-694
1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose 98%
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose

Precaución: for Research Use Only

Formula: C₂₆H₂₂O₈
MDL Number: MFCD00080818
CAS Number: 22224-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR