Ethyl 4-ethynylbenzoate 97%

Supplier: Ambeed
Warning

A190300-1G A190300-250MG A190300-25G A190300-5G
77012-288EA 18 USD
77012-288 77012-290 77012-292 77012-294
Ethyl 4-ethynylbenzoate 97%
Ethyl 4-ethynylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MDL Number: MFCD00168821
CAS Number: 10602-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR