tert-Butyl (4-bromo-2-nitrophenyl)carbamate 95%

Supplier: Ambeed
Warning

A190270-100MG A190270-1G A190270-250MG A190270-5G
76986-074EA 13.8 USD
76986-074 76986-076 76986-078 76986-080
tert-Butyl (4-bromo-2-nitrophenyl)carbamate 95%
tert-Butyl (4-bromo-2-nitrophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrN₂O₄
MW: 317.13 g/mol
MDL Number: MFCD20483775
CAS Number: 327046-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR