3-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester 96%

Supplier: Ambeed
Danger

Synonyms: 3-(Piperidin-1-ylmethyl)phenylboronic acid, pinacol ester, 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperidine

A190256-1G A190256-250MG A190256-5G
77029-794EA 83.82 USD
77029-794 77029-796 77029-798
3-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester 96%
3-(Piperidin-1-ylmethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₂
MW: 301.2314 g/mol
MDL Number: MFCD08435871
CAS Number: 859833-21-9
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR