4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Pyridyl)phenylboronic acid Pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyridine, 4,4,5,5-Tetramethyl-2-[4-(4-pyridyl)phenyl]-1,3,2-dioxaborolane

A189963-100MG A189963-10G A189963-1G A189963-250MG A189963-5G
77029-784EA 10.61 USD
77029-784 77029-786 77029-788 77029-790 77029-792
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 95%
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine

Precaución: for Research Use Only

Formula: C₁₇H₂₀BNO₂
MW: 281.16 g/mol
MDL Number: MFCD12828171
CAS Number: 1009033-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR