5-Methyl-2-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A189858-1G A189858-250MG A189858-5G A189858-25G
77029-778EA 9 USD
77029-778 77029-780 77029-782 77871-954
5-Methyl-2-nitrobenzonitrile 95%
5-Methyl-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C8H6N2O2
MDL Number: MFCD00267507
CAS Number: 64113-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR