Methyl 4-amino-5-methylthiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

A189823-1G A189823-250MG
76986-044EA 38.2 USD
76986-044 76986-046
Methyl 4-amino-5-methylthiophene-3-carboxylate 98%
Methyl 4-amino-5-methylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
MDL Number: MFCD20526415
CAS Number: 81528-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR