Boc-Arg-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-5-guanidinopentanoic acid

A189778-100G A189778-10G A189778-1G A189778-5G
76998-916EA 129 USD
76998-916 76998-918 76998-920 76998-922
Boc-Arg-OH 95%
Boc-Arg-OH

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₄O₄
MDL Number: MFCD00042632
CAS Number: 13726-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR