1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboxamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboximidamide hydrochloride

A189655-1G A189655-25G A189655-5G A189655-100G
76986-038EA 14.86 USD
76986-038 76986-040 76986-042 77683-488
1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboxamidine hydrochloride 97%
1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboxamidine hydrochloride

Precaución: For research use only.

Formula: C₁₄H₁₃ClFN₅
MW: 305.74 g/mol
MDL Number: MFCD18432772
CAS Number: 256499-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR