2-Methyl-4-phenylphenol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-[1,1'-biphenyl]-4-ol

A189644-1G A189644-250MG
77012-248EA 64.72 USD
77012-248 77012-250 77012-252
2-Methyl-4-phenylphenol 98%
2-Methyl-4-phenylphenol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MDL Number: MFCD18313070
CAS Number: 30451-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR