5-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)pyridine

A189261-100MG A189261-1G A189261-250MG A189261-5G
77029-770EA 11 USD
77029-770 77667-578 77667-580 77871-949
5-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester 98%
5-(Trifluoromethyl)-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₃NO₂
MW: 273.0592 g/mol
MDL Number: MFCD12198142
CAS Number: 1084953-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR