2,6-Dibromo-3-pyridinecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dibromopyridine-3-carboxylic acid

A189218-100G A189218-10G A189218-1G A189218-250MG A189218-25G A189218-5G
77029-758EA 934.66 USD
77029-758 77029-760 77029-762 77029-764 77029-766 77029-768
2,6-Dibromo-3-pyridinecarboxylic acid 97%
2,6-Dibromo-3-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MW: 280.90 g/mol
MDL Number: MFCD11846496
CAS Number: 55304-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR