1-(Dimethylamino)-6-methylheptan-3-one 95%

Supplier: Ambeed
Warning

A188816-100MG A188816-1G A188816-250MG A188816-5G
77012-218EA 63 USD
77012-218 77012-220 77012-222 77012-224
1-(Dimethylamino)-6-methylheptan-3-one 95%

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO
MDL Number: MFCD22683883
CAS Number: 107245-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR