6-(Benzyloxy)-3,4-dihydronaphthalen-1(2H)-one 97%

Supplier: Ambeed
Warning

A188693-100MG A188693-10G A188693-1G A188693-250MG A188693-25G A188693-5G
76986-014EA 5.31 USD
76986-014 76986-016 76986-018 76986-020 76986-022 77683-485
6-(Benzyloxy)-3,4-dihydronaphthalen-1(2H)-one 97%
6-(Benzyloxy)-3,4-dihydronaphthalen-1(2H)-one

Precaución: For research use only.

Formula: C₁₇H₁₆O₂
MW: 252.31 g/mol
MDL Number: MFCD12024333
CAS Number: 32263-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR